Gene Transcripts

Gene Symbol
Gdf2
Entrez Gene ID
12165
Gene Name
growth differentiation factor 2
Gene Aliases
Bmp9
Chromosome Location
Chr. 14: 33941039 - 33947198 on Build GRCm38
UniGene ID
Mm.422844
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019506.4 NP_062379.3 2 620
GenBank mRNA AF156890.1 - 2 620
AK147029.1 - 2 621
BC103625.1 - 2 589 40047137
BC103679.1 - 2 589 40047139
BC103680.1 - 2 589 40047138
BC103681.1 - 2 589 40047136

View Full Product Details