Gene Transcripts
- Gene Symbol
- Clcnka
- Entrez Gene ID
- 12733
- Gene Name
- chloride channel Ka
- Gene Aliases
- C75963, CLC-K1, Clcnk1
- Chromosome Location
- Chr. 4: 141384611 - 141398706 on Build GRCm38
- UniGene ID
- Mm.167900
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC037077.1 |
- |
10 |
989 |
|
View Full Product Details