Gene Transcripts

Gene Symbol
Col4a1
Entrez Gene ID
12826
Gene Name
collagen, type IV, alpha 1
Gene Aliases
Bru, Col4a-1, Del(8)44H, Raw, Svc
Chromosome Location
Chr. 8: 11198423 - 11312826 on Build GRCm38
UniGene ID
Mm.738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009931.2 NP_034061.2 32, 33 2859
GenBank mRNA AK146487.1 - 32, 33 2808
AK147284.1 - 32, 33 2781
AK147355.1 - 32, 33 2784
AK147661.1 - 32, 33 2825
AK159130.1 - 32, 33 2819
BC072650.1 - 32, 33 2722 6852661
J04694.1 - 32, 33 2763
X06777.1 - 32, 33 2782

View Full Product Details