Gene Transcripts

Gene Symbol
Cyp3a16
Entrez Gene ID
13114
Gene Name
cytochrome P450, family 3, subfamily a, polypeptide 16
Gene Aliases
Chromosome Location
Chr. 5: 145436309 - 145469737 on Build GRCm38
UniGene ID
Mm.378905
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007820.2 NP_031846.2 11 1184
GenBank mRNA BC113143.1 - 11 1149
D26137.1 - 11 1155

View Full Product Details