Gene Transcripts

Gene Symbol
Efnb1
Entrez Gene ID
13641
Gene Name
ephrin B1
Gene Aliases
Cek5-L, EFL-3, Elk-L, Epl2, Eplg2, LERK-2, Lerk2, Stra1
Chromosome Location
Chr. X: 99136130 - 99148991 on Build GRCm38
UniGene ID
Mm.3374
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK034996.1 - 3 1231
BC006797.1 - 3 1200 2648527
BC018572.1 - 3 1041
BC021656.1 - 3 1213 3602195
U12983.1 - 3 728
Z48781.1 - 3 1191

View Full Product Details