Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008471.2 | NP_032497.1 | 1 | 450 | |
GenBank mRNA | AB472075.1 | - | 1 | 450 | |
BC034561.1 | - | 1 | 443 | 2655016 | |
BC085304.1 | - | 1 | 450 | 6437375 | |
M28698.1 | - | 1 | 445 |