Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008894.2 | NP_032920.2 | 4 | 552 | |
GenBank mRNA | AK013901.1 | - | 4 | 519 | |
AK181645.1 | - | 106 | |||
AK218431.1 | - | 3 | 267 | ||
BC003342.1 | - | 4 | 518 | 3257610 | |
BG078511.2 | - | 4 | 505 | ||
CB205181.1 | - | 1 | 88 | ||
Z72486.1 | - | 4 | 513 |