Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011400.3 | NP_035530.2 | 2, 3 | 334 | |
GenBank mRNA | AK089353.1 | - | 2, 3 | 276 | |
AK155252.1 | - | 2, 3 | 276 | ||
AK170425.1 | - | 2, 3 | 276 | ||
AK170873.1 | - | 2, 3 | 276 | ||
BC055340.1 | - | 2, 3 | 303 | 6808315 | |
BY233228.1 | - | 2, 3 | 334 | ||
M22998.1 | - | 2, 3 | 238 | ||
M23384.1 | - | 2, 3 | 313 |