Gene Transcripts

Gene Symbol
Ttbk1
Entrez Gene ID
106763
Gene Name
tau tubulin kinase 1
Gene Aliases
AU017937, AW048023, C330008L01Rik
Chromosome Location
Chr. 17: 46442448 - 46487675 on Build GRCm38
UniGene ID
Mm.297613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162864.1 NP_001156336.1 7 928

View Full Product Details