Gene Transcripts

Gene Symbol
Mrgpra6
Entrez Gene ID
381886
Gene Name
MAS-related GPR, member A6
Gene Aliases
MrgA6
Chromosome Location
Chr. 7: 47185766 - 47189355 on Build GRCm38
UniGene ID
Mm.377894
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC132198.1 - 1 543
BC132204.1 - 1 543 40130481

View Full Product Details