Gene Transcripts

Gene Symbol
Tap1
Entrez Gene ID
21354
Gene Name
transporter 1, ATP-binding cassette, sub-family B (MDR/TAP)
Gene Aliases
ABC17, APT1, Abcb2, Ham-1, Ham1, MTP1, PSF1, RING4, TAP, Tap-1, Y3
Chromosome Location
Chr. 17: 34187556 - 34197225 on Build GRCm38
UniGene ID
Mm.482076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161730.1 NP_001155202.1 12 2250
NM_013683.2 NP_038711.2 11 2334
GenBank mRNA AK040919.1 - 9 3297
AK151481.1 - 11 2089
AK152953.1 - 11 2034
AK153890.1 - 11 2338
AK162731.1 - 11 2134
AK166046.1 - 11 2337
AK167485.1 - 11 2336
AK171363.1 - 12 2005
BC013802.1 - 11 1745 3257566
BC024897.1 - 11 1857 3257734
BC031584.1 - 7 2469
KJ650227.1 - 11 2010
M55637.1 - 11 1726
U60018.1 - 11 2010
U60019.1 - 11 2010
U60020.1 - 11 2010
U60021.1 - 11 2010
U60022.1 - 11 2010
U60023.1 - 11 2010
X59615.1 - 11 1569

View Full Product Details