Gene Transcripts

Gene Symbol
Vwa3b
Entrez Gene ID
70853
Gene Name
von Willebrand factor A domain containing 3B
Gene Aliases
4921511C04Rik, A230074B11Rik, Gm555
Chromosome Location
Chr. 1: 37026591 - 37187608 on Build GRCm38
UniGene ID
Mm.148848
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK038906.1 - 2, 3 344
AK050189.1 - 2, 3 210
AK076981.1 - 2, 3 338
BC049712.1 - 2, 3 355
BC147491.1 - 2, 3 204 9087438

View Full Product Details