Gene Transcripts
- Gene Symbol
- Tpi1
- Entrez Gene ID
- 21991
- Gene Name
- triosephosphate isomerase 1
- Gene Aliases
- AI255506, Tpi, Tpi-1
- Chromosome Location
- Chr. 6: 124810592 - 124814296 on Build GRCm38
- UniGene ID
- Mm.4222
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009415.2 |
NP_033441.2
|
3, 4 |
495 |
|
GenBank mRNA |
AK002270.1 |
- |
3, 4 |
357 |
|
AK010808.1 |
- |
3, 4 |
365 |
|
AK146013.1 |
- |
3, 4 |
354 |
|
AK149768.1 |
- |
3, 4 |
359 |
|
AK150735.1 |
- |
3, 4 |
356 |
|
AK159741.1 |
- |
3, 4 |
359 |
|
AK168060.1 |
- |
3, 4 |
357 |
|
AK168446.1 |
- |
3, 4 |
359 |
|
AK168756.1 |
- |
3, 4 |
358 |
|
BC004024.1 |
- |
3, 4 |
343 |
|
BC046761.1 |
- |
3, 4 |
346 |
5693696 |
BU945752.1 |
- |
3, 4 |
500 |
|
X53333.1 |
- |
3, 4 |
357 |
|
View Full Product Details