Gene Transcripts

Gene Symbol
Pebp1
Entrez Gene ID
23980
Gene Name
phosphatidylethanolamine binding protein 1
Gene Aliases
HCNP, Pbp, Pbp1, Pbpr, Rkip
Chromosome Location
Chr. 5: 117282651 - 117287564 on Build GRCm38
UniGene ID
Mm.195898
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018858.2 NP_061346.2 2 251
GenBank mRNA AB046417.1 - 2 253
AF300422.1 - 2 230
AK008037.1 - 267
AK088212.1 - 2 256
AK143897.1 - 2 283
AK149815.1 - 2 267
AK159842.1 - 2 256
AK182328.1 - 2 141
BC008169.1 - 2 285 3257403
BC083063.1 - 2 251 5362125
BC089332.1 - 2 234 30359654
U43206.1 - 2 249

View Full Product Details