Gene Transcripts

Gene Symbol
Zbtb39
Entrez Gene ID
320080
Gene Name
zinc finger and BTB domain containing 39
Gene Aliases
7030401O21Rik, mKIAA0352
Chromosome Location
Chr. 10: 127739538 - 127747349 on Build GRCm38
UniGene ID
Mm.27258
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198035.1 NP_932152.1 2 381
GenBank mRNA AK135409.1 - 2 373
AK155700.1 - 2 357
AK172936.1 - 1 134
BC058661.1 - 2 381 6409441
BC079569.1 - 2 277 30536951

View Full Product Details