Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001038710.1 | NP_001033799.1 | 4 | 820 | |
NM_016711.3 | NP_057920.2 | 4 | 756 | ||
GenBank mRNA | AF177170.1 | - | 4 | 705 | |
AK015263.1 | - | 4 | 801 | ||
AK015719.1 | - | 4 | 856 | ||
AK017072.1 | - | 4 | 804 | ||
AK018223.1 | - | 4 | 553 | ||
AK076562.1 | - | 4 | 770 | ||
AK079077.1 | - | 4 | 600 | ||
AK147581.1 | - | 4 | 752 | ||
AK164178.1 | - | 4 | 602 | ||
BC052227.1 | - | 4 | 820 | ||
BC061124.1 | - | 4 | 586 | 6531518 |