Gene Transcripts

Gene Symbol
Rgs7bp
Entrez Gene ID
52882
Gene Name
regulator of G-protein signalling 7 binding protein
Gene Aliases
A930030I01Rik, AI842293, D13Bwg1146e, R7bp
Chromosome Location
Chr. 13: 104945904 - 105054930 on Build GRCm38
UniGene ID
Mm.100348
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029879.2 NP_084155.2 3 1062
GenBank mRNA AK020910.1 - 3 659
AK028323.1 - 3 439
AK032138.1 - 3 1062
AK036240.1 - 3 969
AK046733.1 - 3 691
AK137417.1 - 3 936
BC118957.2 - 3 594 40093423
DQ104214.1 - 3 352

View Full Product Details