Gene Transcripts

Gene Symbol
Mybph
Entrez Gene ID
53311
Gene Name
myosin binding protein H
Gene Aliases
AI385643, MyBP-H
Chromosome Location
Chr. 1: 134193417 - 134201234 on Build GRCm38
UniGene ID
Mm.379067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016749.2 NP_058029.2 1 264
GenBank mRNA AA673795.1 - 1 276
BC118050.1 - 1 231 40106744
U68267.1 - 1 258

View Full Product Details