Gene Transcripts

Gene Symbol
Sac3d1
Entrez Gene ID
66406
Gene Name
SAC3 domain containing 1
Gene Aliases
2410004C24Rik, AU015683, shd1
Chromosome Location
Chr. 19: 6116004 - 6118586 on Build GRCm38
UniGene ID
Mm.303924
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133678.3 NP_598439.3 1 46
GenBank mRNA AJ131957.2 - 1 46
BC144812.1 - 1 110 9053360
BC145789.1 - 1 110 40131092

View Full Product Details