Gene Transcripts
- Gene Symbol
- Ydjc
- Entrez Gene ID
- 69101
- Gene Name
- YdjC homolog (bacterial)
- Gene Aliases
- 1810015A11Rik, 4930521M19Rik
- Chromosome Location
- Chr. 16: 17146882 - 17148857 on Build GRCm38
- UniGene ID
- Mm.101656
- Species
- Mus musculus
- Gene Symbol
- Ccdc116
- Entrez Gene ID
- 76872
- Gene Name
- coiled-coil domain containing 116
- Gene Aliases
- 4930432J16Rik
- Chromosome Location
- Chr. 16: 17139064 - 17147146 on Build GRCm38
- UniGene ID
- Mm.101656
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK141314.1 |
- |
1 |
259 |
|
View Full Product Details