Gene Transcripts

Gene Symbol
Cdc16
Entrez Gene ID
69957
Gene Name
CDC16 cell division cycle 16
Gene Aliases
2700071J12Rik, 2810431D22Rik, APC6
Chromosome Location
Chr. 8: 13757646 - 13781938 on Build GRCm38
UniGene ID
Mm.182412
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027276.2 NP_081552.2 12, 13 1224
GenBank mRNA AK013213.1 - 1, 2 158
AK147938.1 - 12, 13 1239
AK167965.1 - 12, 13 1239
AK168978.1 - 12, 13 1211
AK176938.1 - 2, 3 199
AK190066.1 - 1, 2 125
AK213282.1 - 1, 2 125
AK215012.1 - 1, 2 125
AK219967.1 - 1, 2 125
BC026606.1 - 12, 13 1224 4976424

View Full Product Details