Gene Transcripts

Gene Symbol
Tmco6
Entrez Gene ID
71983
Gene Name
transmembrane and coiled-coil domains 6
Gene Aliases
2410015B03Rik
Chromosome Location
Chr. 18: 36735018 - 36742400 on Build GRCm38
UniGene ID
Mm.135188
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028036.3 NP_082312.3 11 1408
GenBank mRNA AK010487.1 - 11 1333
BC005637.1 - 11 1410 3156734
BF468256.2 - 3 276

View Full Product Details