Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031389.2 | NP_113566.2 | 8 | 2688 | |
GenBank mRNA | AF285581.1 | - | 6 | 1956 | |
AK133284.1 | - | 8 | 2690 | ||
AK166817.1 | - | 8 | 2669 | ||
AK166869.1 | - | 8 | 2914 | ||
AY596194.1 | - | 8 | 2688 | ||
BC120833.1 | - | 6 | 1949 | 40129756 |