Gene Transcripts

Gene Symbol
Msh3
Entrez Gene ID
17686
Gene Name
mutS homolog 3 (E. coli)
Gene Aliases
D13Em1, Rep-3, Rep3
Chromosome Location
Chr. 13: 92211879 - 92355003 on Build GRCm38
UniGene ID
Mm.343101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010829.2 NP_034959.2 4 677
GenBank mRNA AK011426.1 - 4 661
AK040616.1 - 3 462
AK078467.1 - 4 628
AK083388.1 - 4 582
AK133907.1 - 4 670
AK161089.1 - 4 672
M24919.1 - 4 960
M80360.1 - 4 674

View Full Product Details