Gene Transcripts
- Gene Symbol
- Tdpoz3
- Entrez Gene ID
- 399674
- Gene Name
- TD and POZ domain containing 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 93826020 - 93827117 on Build GRCm38
- UniGene ID
- Mm.377923
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_207271.2 |
NP_997154.2
|
1 |
514 |
|
View Full Product Details