Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019821.2 | NP_062795.2 | 5 | 684 | |
GenBank mRNA | AK003218.1 | - | 1 | 151 | |
AK157313.1 | - | 5 | 671 | ||
BC016584.1 | - | 5 | 684 | 3157800 | |
BC053729.1 | - | 5 | 627 | 30065192 |