Gene Transcripts

Gene Symbol
Katnal2
Entrez Gene ID
71206
Gene Name
katanin p60 subunit A-like 2
Gene Aliases
3110023G01Rik, 4933439B08Rik
Chromosome Location
Chr. 18: 76977147 - 77048225 on Build GRCm38
UniGene ID
Mm.35803
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027721.2 NP_081997.1 9 829
GenBank mRNA AK014074.1 - 10 940
AK017114.1 - 9 830
BC056963.1 - 9 776

View Full Product Details