Gene Transcripts

Gene Symbol
Fancf
Entrez Gene ID
100040608
Gene Name
Fanconi anemia, complementation group F
Gene Aliases
A730016A17
Chromosome Location
Chr. 7: 51860577 - 51862267 on Build GRCm38
UniGene ID
Mm.322643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115087.1 NP_001108559.1 1 1012
GenBank mRNA AK139575.1 - 1 483

View Full Product Details