Gene Transcripts
- Gene Symbol
- Zfp74
- Entrez Gene ID
- 72723
- Gene Name
- zinc finger protein 74
- Gene Aliases
- 2810054M15Rik, KRAB8, Zfp66, Znf569, mszf21, mszf77
- Chromosome Location
- Chr. 7: 29931711 - 29955843 on Build GRCm38
- UniGene ID
- Mm.371648
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK051279.1 |
- |
1 |
117 |
|
BC034883.1 |
- |
1 |
113 |
|
- Gene Symbol
- C230062I16Rik
- Entrez Gene ID
- 625421
- Gene Name
- RIKEN cDNA C230062I16 gene
- Gene Aliases
- Chromosome Location
- Chr. 7: 29956984 - 29969764 on Build GRCm38
- UniGene ID
- Mm.464976
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK082547.1 |
- |
|
327 |
|
View Full Product Details