Gene Transcripts

Gene Symbol
Cyp4f15
Entrez Gene ID
106648
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 15
Gene Aliases
AI787289, Cyp4f-15
Chromosome Location
Chr. 17: 32685601 - 32703352 on Build GRCm38
UniGene ID
Mm.26539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134127.1 NP_598888.1 2 270
GenBank mRNA AF233645.1 - 2 270
BC021377.1 - 2 261 5050617

View Full Product Details