Gene Transcripts
- Gene Symbol
- Tnfaip8
- Entrez Gene ID
- 106869
- Gene Name
- tumor necrosis factor, alpha-induced protein 8
- Gene Aliases
- AA987150, E130304C20Rik, ENSMUSG00000073567, Gg2-1, Gm10539, Nded, Ssc-2
- Chromosome Location
- Chr. 18: 49979427 - 50093229 on Build GRCm38
- UniGene ID
- Mm.27740
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_134131.2 |
NP_598892.2
|
1, 2 |
345 |
|
GenBank mRNA |
AK087478.1 |
- |
1, 2 |
263 |
|
AK090316.1 |
- |
1, 2 |
155 |
|
AK090350.1 |
- |
1, 2 |
231 |
|
AK149901.1 |
- |
1, 2 |
121 |
|
AK150445.1 |
- |
1, 2 |
117 |
|
AK150965.1 |
- |
1, 2 |
127 |
|
AK151622.1 |
- |
1, 2 |
117 |
|
AK152426.1 |
- |
1, 2 |
117 |
|
BC009090.1 |
- |
1, 2 |
107 |
3965693 |
BY108131.1 |
- |
1, 2 |
346 |
|
View Full Product Details