Gene Transcripts

Gene Symbol
Ly6g5c
Entrez Gene ID
114652
Gene Name
lymphocyte antigen 6 complex, locus G5C
Gene Aliases
G5c, NG33
Chromosome Location
Chr. 17: 35108300 - 35111953 on Build GRCm38
UniGene ID
Mm.222643
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_148947.1 NP_683749.1 2 171
GenBank mRNA AJ315552.1 - 2 171
BC111447.1 - 2 170
BC145966.1 - 2 192 40131269
BC145968.1 - 2 192 40131271

View Full Product Details