Gene Transcripts

Gene Symbol
Prosc
Entrez Gene ID
114863
Gene Name
proline synthetase co-transcribed
Gene Aliases
1700024N20Rik, 2200002F22Rik
Chromosome Location
Chr. 8: 27042555 - 27056132 on Build GRCm38
UniGene ID
Mm.277501
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039077.2 NP_001034166.1 5 397
NM_001039078.2 NP_001034167.1 4 361
NM_054057.4 NP_473398.1 5 397
GenBank mRNA AK032308.1 - 5 347
AK049905.1 - 4 3530
AK134549.1 - 5 330
AK147670.1 - 5 379
AK160172.1 - 5 351
AK211128.1 - 1 19
AK213501.1 - 2 35
AK218333.1 - 2 35
BC047992.1 - 4 318 6504579
BC061045.1 - 5 391 6534037

View Full Product Details