Gene Transcripts

Gene Symbol
Bmp8b
Entrez Gene ID
12164
Gene Name
bone morphogenetic protein 8b
Gene Aliases
Op3
Chromosome Location
Chr. 4: 123104307 - 123126094 on Build GRCm38
UniGene ID
Mm.439764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007559.4 NP_031585.2 4 984
GenBank mRNA AK038590.1 - 206
AK082895.1 - 4 985
BC137890.1 - 4 854 8860909
U39545.1 - 4 984

View Full Product Details