Gene Transcripts

Gene Symbol
Mia
Entrez Gene ID
12587
Gene Name
melanoma inhibitory activity
Gene Aliases
CD-RAP, Cdrap, Mia1
Chromosome Location
Chr. 7: 27179741 - 27181149 on Build GRCm38
UniGene ID
Mm.262447
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019394.3 NP_062267.2 1, 2 221
GenBank mRNA AK135151.1 - 1, 2 200
AW047492.1 - 3, 4 359
BC009815.1 - 1, 2 190
BE864464.1 - 1, 2 221
CK794645.1 - 1, 2 215
X94322.1 - 1, 2 226

View Full Product Details