Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001042634.2 | NP_001036099.1 | 2 | 305 | |
GenBank mRNA | AK031218.1 | - | 2 | 239 | |
AK076199.1 | - | 2 | 240 | ||
AK135765.1 | - | 2 | 240 | ||
BC062887.1 | - | 257 | |||
BC145905.1 | - | 2 | 208 | ||
BC145911.1 | - | 2 | 208 | ||
BY784955.1 | - | 2 | 306 | ||
L29221.1 | - | 2 | 221 | ||
M38381.1 | - | 2 | 221 | ||
U11054.1 | - | 2 | 205 | ||
U21209.1 | - | 2 | 221 | ||
X57186.1 | - | 2 | 205 |