Gene Transcripts
- Gene Symbol
- Asap1
- Entrez Gene ID
- 13196
- Gene Name
- ArfGAP with SH3 domain, ankyrin repeat and PH domain1
- Gene Aliases
- AV239055, DEF-1, Ddef1, PAP, mKIAA1249, s19
- Chromosome Location
- Chr. 15: 64086840 - 64382919 on Build GRCm38
- UniGene ID
- Mm.277236
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276461.1 |
NP_001263390.1
|
3, 4 |
410 |
|
NM_001276462.1 |
NP_001263391.1
|
1, 2 |
319 |
|
NM_001276463.1 |
NP_001263392.1
|
1, 2 |
319 |
|
NM_001276467.1 |
NP_001263396.1
|
3, 4 |
410 |
|
NM_010026.3 |
NP_034156.2
|
3, 4 |
410 |
|
GenBank mRNA |
AF075461.1 |
- |
3, 4 |
229 |
|
AF075462.1 |
- |
3, 4 |
229 |
|
AK122477.1 |
- |
1, 2 |
14 |
|
AK132979.1 |
- |
1, 2 |
149 |
|
AK146545.1 |
- |
3, 4 |
248 |
|
AK147689.1 |
- |
1, 2 |
320 |
|
AK161586.1 |
- |
|
320 |
|
BC048818.1 |
- |
1, 2 |
285 |
|
BC094581.1 |
- |
3, 4 |
410 |
30549241 |
CB527210.1 |
- |
1, 2 |
113 |
|
CF735731.1 |
- |
3, 4 |
410 |
|
View Full Product Details