Gene Transcripts

Gene Symbol
Emx1
Entrez Gene ID
13796
Gene Name
empty spiracles homeobox 1
Gene Aliases
Chromosome Location
Chr. 6: 85187931 - 85204463 on Build GRCm38
UniGene ID
Mm.391473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010131.2 NP_034261.1 3 720
GenBank mRNA AB221645.1 - 3 708
AK143888.1 - 2 807
BC104322.2 - 1 345
BC104323.2 - 1 345

View Full Product Details