Gene Transcripts

Gene Symbol
Gstm6
Entrez Gene ID
14867
Gene Name
glutathione S-transferase, mu 6
Gene Aliases
Chromosome Location
Chr. 3: 107938847 - 107953491 on Build GRCm38
UniGene ID
Mm.458154
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008184.3 NP_032210.3 5 449
GenBank mRNA AJ000413.1 - 5 333
AK015265.1 - 5 435
BC031818.1 - 5 412 4196506
CB573365.1 - 5 452

View Full Product Details