Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008266.5 | NP_032292.3 | 1 | 343 | |
GenBank mRNA | AK017686.1 | - | 1 | 344 | |
BC103597.1 | - | 1 | 340 | 40046066 | |
BC103598.1 | - | 1 | 340 | 40046076 | |
BC103605.1 | - | 1 | 340 | 40046069 | |
BC103606.1 | - | 1 | 340 | 40046068 |