Gene Transcripts

Gene Symbol
Scrt1
Entrez Gene ID
170729
Gene Name
scratch homolog 1, zinc finger protein (Drosophila)
Gene Aliases
Chromosome Location
Chr. 15: 76516203 - 76522129 on Build GRCm38
UniGene ID
Mm.218747
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130893.3 NP_570963.1 2 629
GenBank mRNA BC057562.1 - 2 402
BC089316.1 - 2 413
CB525529.1 - 2 629

View Full Product Details