Gene Transcripts

Gene Symbol
Crb1
Entrez Gene ID
170788
Gene Name
crumbs family member 1, photoreceptor morphogenesis associated
Gene Aliases
7530426H14Rik, A930008G09Rik
Chromosome Location
Chr. 1: 139197546 - 139379118 on Build GRCm38
UniGene ID
Mm.95700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133239.2 NP_573502.2 4 1122
GenBank mRNA AF406641.1 - 4 1087
AF492496.1 - 4 1146
AY450552.1 - 4 1087

View Full Product Details