Gene Transcripts

Gene Symbol
Ptger2
Entrez Gene ID
19217
Gene Name
prostaglandin E receptor 2 (subtype EP2)
Gene Aliases
EP2, Ptgerep2
Chromosome Location
Chr. 14: 44988111 - 45003820 on Build GRCm38
UniGene ID
Mm.4630
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008964.4 NP_032990.1 1 1643
GenBank mRNA AK036474.1 - 1 1088
AK036531.1 - 1 1561
AK054132.1 - 1 1164
AK086904.1 - 1 1558
AK136577.1 - 1 656
AK137515.1 - 1 853
BC005440.1 - 1 1151 3487087
D50589.1 - 1 934

View Full Product Details