Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290283.1 | NP_001277212.1 | 1 | 169 | |
NM_183139.2 | NP_898962.2 | 1 | 169 | ||
GenBank mRNA | AK077214.1 | - | 1 | 131 | |
AK139586.1 | - | 1 | 157 | ||
BC119245.1 | - | 1 | 169 | 8733994 | |
BC145052.1 | - | 1 | 169 |