Gene Transcripts
- Gene Symbol
- Rit1
- Entrez Gene ID
- 19769
- Gene Name
- Ras-like without CAAX 1
- Gene Aliases
- RIBB, ROC1, Rit
- Chromosome Location
- Chr. 3: 88716840 - 88731048 on Build GRCm38
- UniGene ID
- Mm.4009
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_009069.4 |
NP_033095.1
|
2, 3 |
243 |
|
GenBank mRNA |
AK005357.1 |
- |
2, 3 |
204 |
|
AK014126.1 |
- |
2, 3 |
234 |
|
AK018785.1 |
- |
2, 3 |
191 |
|
AK164196.1 |
- |
2, 3 |
234 |
|
AK168254.1 |
- |
2, 3 |
202 |
|
AK186691.1 |
- |
1, 2 |
14 |
|
AK205825.1 |
- |
1, 2 |
14 |
|
AK208119.1 |
- |
1, 2 |
14 |
|
BC012694.1 |
- |
2, 3 |
203 |
4023732 |
BY284832.1 |
- |
2, 3 |
246 |
|
U71205.1 |
- |
2, 3 |
180 |
|
View Full Product Details