Gene Transcripts

Gene Symbol
Uri1
Entrez Gene ID
19777
Gene Name
URI1, prefoldin-like chaperone
Gene Aliases
C80913, NNX3, Rmp
Chromosome Location
Chr. 7: 37959992 - 38023575 on Build GRCm38
UniGene ID
Mm.387220
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011274.5 NP_035404.4 8 1166
GenBank mRNA AF091096.1 - 6 688
AK052034.1 - 3 347
AK133383.1 - 8 841
AK155994.1 - 8 1140
AK162285.1 - 3 340
AK165045.1 - 1 118
AK166566.1 - 8 1157
BC023029.1 - 8 826 5374731

View Full Product Details