Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009139.3 | NP_033165.1 | 4 | 462 | |
GenBank mRNA | AF128188.1 | - | 4 | 358 | |
AF128189.1 | - | 4 | 358 | ||
AF128190.1 | - | 4 | 358 | ||
AF128191.1 | - | 4 | 358 | ||
AF128192.1 | - | 4 | 358 | ||
AK002697.1 | - | 4 | 365 | ||
AK008547.1 | - | 4 | 366 | ||
AK150336.1 | - | 4 | 365 | ||
AK154553.1 | - | 4 | 366 | ||
BC002073.1 | - | 4 | 355 | 3492808 | |
M58004.1 | - | 4 | 365 |