Gene Transcripts
- Gene Symbol
- Slc31a2
- Entrez Gene ID
- 20530
- Gene Name
- solute carrier family 31, member 2
- Gene Aliases
- AI604396, CTR2
- Chromosome Location
- Chr. 4: 62280393 - 62298412 on Build GRCm38
- UniGene ID
- Mm.292539
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290518.1 |
NP_001277447.1
|
3, 4 |
354 |
|
NM_025286.3 |
NP_079562.1
|
3, 4 |
392 |
|
GenBank mRNA |
AK003269.1 |
- |
3, 4 |
327 |
|
AK007579.1 |
- |
3, 4 |
349 |
|
AK008473.1 |
- |
3, 4 |
376 |
|
AK015878.1 |
- |
3, 4 |
355 |
|
AK151612.1 |
- |
3, 4 |
322 |
|
AK152812.1 |
- |
3, 4 |
349 |
|
BB853212.1 |
- |
3, 4 |
392 |
|
BC029183.1 |
- |
3, 4 |
283 |
4502064 |
BC099447.1 |
- |
3, 4 |
358 |
30715212 |
View Full Product Details