Gene Transcripts

Gene Symbol
Snrpe
Entrez Gene ID
20643
Gene Name
small nuclear ribonucleoprotein E
Gene Aliases
AL022645, C76690, SME
Chromosome Location
Chr. 1: 133603871 - 133610280 on Build GRCm38
UniGene ID
Mm.249110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009227.3 NP_033253.1 3, 4 198
GenBank mRNA AK019462.1 - 3, 4 182
AW492281.1 - 2, 3 307
BC008262.1 - 3, 4 174 2650413
BC051207.1 - 3, 4 156 1396204
BC055765.1 - 3, 4 140 6407944
BY168551.1 - 3, 4 198
X65704.1 - 3, 4 102

View Full Product Details