Gene Transcripts
- Gene Symbol
- Hectd1
- Entrez Gene ID
- 207304
- Gene Name
- HECT domain containing 1
- Gene Aliases
- A630086P08Rik, AI844876, b2b327Clo, opm
- Chromosome Location
- Chr. 12: 51743721 - 51829605 on Build GRCm38
- UniGene ID
- Mm.249391
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_144788.2 |
NP_659037.2
|
32 |
6147 |
|
GenBank mRNA |
AK173106.1 |
- |
12 |
2676 |
|
View Full Product Details